MAN238931765 Manchester Organics

3,4-Difluoro-2-methoxybenzoic acid

  • MDL number MFCD03428593
  • Molecular Weight : 188.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B21550

Safety Information

MSDS https://www.manchesterorganics.com/B21550
Availability details for
Quantity
  • Availability
  • Add to Cart