MAN238931770 Manchester Organics

3-Chloro-4-nitroaniline

  • MDL number MFCD00085922
  • Molecular Weight : 172.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G21580

Safety Information

MSDS https://www.manchesterorganics.com/G21580
Availability details for
Quantity
  • Availability
  • Add to Cart