MAN238931802 Manchester Organics

3-Phenylpyridine

  • MDL number MFCD00006380
  • Cas Number : 1008-88-4
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-phenylpyridine
Inchi Code 1S/C11H9N/c1-2-5-10(6-3-1)11-7-4-8-12-9-11/h1-9H
Inchi Key HJKGBRPNSJADMB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R21540
Availability details for
Quantity
  • Availability
  • Add to Cart