MAN238931804 Manchester Organics

2-Fluoro-5-(trifluoromethoxy)benzonitrile

  • MDL number MFCD04115894
  • Molecular Weight : 205.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F21295
Availability details for
Quantity
  • Availability
  • Add to Cart