MAN238931832 Manchester Organics

2-Amino-3-methylpyrazine

  • MDL number MFCD08705770
  • Cas Number : 19838-08-5
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J21557
Availability details for
Quantity
  • Availability
  • Add to Cart