MAN238931839 Manchester Organics

Amino(2-bromophenyl)acetic acid

  • MDL number MFCD02662413
  • Cas Number : 254762-66-8
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N21561
Availability details for
Quantity
  • Availability
  • Add to Cart