MAN326366553 Manchester Organics

2-Methyl-4-(trifluoromethyl)pyridine

  • MDL number MFCD13185742
  • Molecular Weight : 161.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S14631
Availability details for
Quantity
  • Availability
  • Add to Cart