MAN326366572 Manchester Organics

4-(1,3,4-Oxadiazol-2-yl)piperidine

  • MDL number MFCD11042734
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H23427

Safety Information

MSDS https://www.manchesterorganics.com/H23427
Availability details for
Quantity
  • Availability
  • Add to Cart