MAN326366580 Manchester Organics

2-Amino-5-hydroxypyridine hydrochloride

  • MDL number MFCD11506259
  • Molecular Weight : 146.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R20688

Safety Information

MSDS https://www.manchesterorganics.com/R20688
Availability details for
Quantity
  • Availability
  • Add to Cart