MAN326366609 Manchester Organics

1-Phenyl-cyclobutanecarbonitrile

  • MDL number MFCD00019258
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G15403

Safety Information

MSDS https://www.manchesterorganics.com/G15403
Availability details for
Quantity
  • Availability
  • Add to Cart