MAN326366617 Manchester Organics

5-Bromo-3-hydroxy-3H-isobenzofuran-1-one

  • MDL number MFCD09836746
  • Molecular Weight : 229.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M15337
Availability details for
Quantity
  • Availability
  • Add to Cart