MAN326366625 Manchester Organics

3-Amino-5-bromo-2-methylpyridine

  • MDL number MFCD09031418
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A21597
Availability details for
Quantity
  • Availability
  • Add to Cart