MAN326366626 Manchester Organics

7-Aminoquinoline

  • MDL number MFCD01646243
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B21598
Availability details for
Quantity
  • Availability
  • Add to Cart