MAN326366632 Manchester Organics

5-(4-Fluoro-benzyl)-thiazol-2-ylamine

  • MDL number MFCD02214714
  • Cas Number : 315241-39-5
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-fluorobenzyl)-1,3-thiazol-2-amine
Inchi Code 1S/C10H9FN2S/c11-8-3-1-7(2-4-8)5-9-6-13-10(12)14-9/h1-4,6H,5H2,(H2,12,13)
Inchi Key KIZWUMGELVDUQL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/B11019
Availability details for
Quantity
  • Availability
  • Add to Cart