MAN326366640 Manchester Organics

N-alpha-Formyl-d-leucine

  • MDL number MFCD08275788
  • Cas Number : 44978-39-4
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/C11020
Availability details for
Quantity
  • Availability
  • Add to Cart