MAN326366649 Manchester Organics

Benzoylfluoride

  • MDL number MFCD00000364
  • Molecular Weight : 124.1124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K21606

Safety Information

MSDS https://www.manchesterorganics.com/K21606
Availability details for
Quantity
  • Availability
  • Add to Cart