MAN326366658 Manchester Organics

2-Bromo-4-cyanopyridine

  • MDL number MFCD07367879
  • Molecular Weight : 183.005

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V21615
Availability details for
Quantity
  • Availability
  • Add to Cart