MAN326366683 Manchester Organics

2-Ethoxypyridine

  • MDL number MFCD00023464
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y22115

Safety Information

MSDS https://www.manchesterorganics.com/Y22115
Availability details for
Quantity
  • Availability
  • Add to Cart