MAN326366689 Manchester Organics

4-Fluorobenzenesulfinic acid sodium salt

  • MDL number MFCD03093775
  • Molecular Weight : 182.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A21668

Safety Information

MSDS https://www.manchesterorganics.com/A21668
Availability details for
Quantity
  • Availability
  • Add to Cart