MAN326366694 Manchester Organics

3-Hydroxy-5-methylpyridine

  • MDL number MFCD00661297
  • Cas Number : 42732-49-0
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G21651
Availability details for
Quantity
  • Availability
  • Add to Cart