MAN326366702 Manchester Organics

3-Morpholine carboxylic acid hydrochloride

  • MDL number MFCD01318649
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S21848

Safety Information

MSDS https://www.manchesterorganics.com/S21848
Availability details for
Quantity
  • Availability
  • Add to Cart