MAN326366705 Manchester Organics

N-(3-Chlorophenyl)thiourea

  • MDL number MFCD00022165
  • Molecular Weight : 186.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J23309

Safety Information

MSDS https://www.manchesterorganics.com/J23309
Availability details for
Quantity
  • Availability
  • Add to Cart