MAN326366739 Manchester Organics

3-Methoxybenzophenone

  • MDL number MFCD01319936
  • Molecular Weight : 212.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F21767

Safety Information

MSDS https://www.manchesterorganics.com/F21767
Availability details for
Quantity
  • Availability
  • Add to Cart