MAN326366742 Manchester Organics

(+/-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

  • MDL number MFCD07771983
  • Molecular Weight : 169.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J21770

Safety Information

MSDS https://www.manchesterorganics.com/J21770
Availability details for
Quantity
  • Availability
  • Add to Cart