MAN326366744 Manchester Organics

2,2,2'2'-(5-Methyl-1,3-phenylene)-dimethyl-propionitrile

  • MDL number MFCD08062446
  • Cas Number : 120511-72-0
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/L21701
Availability details for
Quantity
  • Availability
  • Add to Cart