MAN326366755 Manchester Organics

2-Bromomethyl-1,3,5-trimethylbenzene

  • MDL number MFCD09056829
  • Cas Number : 4761-00-6
  • Molecular Weight : 213.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(bromomethyl)-1,3,5-trimethylbenzene
Inchi Code 1S/C10H13Br/c1-7-4-8(2)10(6-11)9(3)5-7/h4-5H,6H2,1-3H3
Inchi Key HIQNEJUMZTWMLW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z21713
Availability details for
Quantity
  • Availability
  • Add to Cart