MAN326366765 Manchester Organics

3-Chloro-4-nitrobenzenesulfonyl chloride

  • MDL number MFCD04973767
  • Cas Number : 64835-30-9
  • Molecular Weight : 256.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/H21723
Availability details for
Quantity
  • Availability
  • Add to Cart