MAN326366791 Manchester Organics

3-Bromo-4-methoxybenzyl alcohol

  • MDL number MFCD01861398
  • Molecular Weight : 217.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L21843

Safety Information

MSDS https://www.manchesterorganics.com/L21843
Availability details for
Quantity
  • Availability
  • Add to Cart