MAN326366818 Manchester Organics

Methyl(4-cyanophenyl)acetate

  • MDL number MFCD02181140
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R21943

Safety Information

MSDS https://www.manchesterorganics.com/R21943
Availability details for
Quantity
  • Availability
  • Add to Cart