MAN326366851 Manchester Organics

1,4-Difluoro-2,3-dimethylbenzene

  • MDL number MFCD04973769
  • Cas Number : 1736-90-9
  • Molecular Weight : 142.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/A21810
Availability details for
Quantity
  • Availability
  • Add to Cart