MAN326366854 Manchester Organics

5-Chloro-2-fluorophenylacetic acid

  • MDL number MFCD01631439
  • Molecular Weight : 188.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E21885

Safety Information

MSDS https://www.manchesterorganics.com/E21885
Availability details for
Quantity
  • Availability
  • Add to Cart