MAN326366861 Manchester Organics

3-Fluoro-4-methoxybenzoyl chloride

  • MDL number MFCD03428589
  • Molecular Weight : 188.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M21940

Safety Information

MSDS https://www.manchesterorganics.com/M21940
Availability details for
Quantity
  • Availability
  • Add to Cart