MAN326366862 Manchester Organics

1-tert-Butyl 3-methyl-5-hydroxypiperidine-1,3-dicarboxylate

  • MDL number MFCD11656770
  • Cas Number : 1095010-47-1
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-tert-butyl 3-methyl 5-hydroxy-1,3-piperidinedicarboxylate
Inchi Code 1S/C12H21NO5/c1-12(2,3)18-11(16)13-6-8(10(15)17-4)5-9(14)7-13/h8-9,14H,5-7H2,1-4H3
Inchi Key DFZMPKMSTNKZKL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/N21822
Availability details for
Quantity
  • Availability
  • Add to Cart