MAN326366863 Manchester Organics

1,2-Bis-(trifluoromethyl)benzene

  • MDL number MFCD03094218
  • Molecular Weight : 214.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D14500
Availability details for
Quantity
  • Availability
  • Add to Cart