MAN326366867 Manchester Organics

3,5-Dimethylbenzotrifluoride

  • MDL number MFCD12026302
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H14575

Safety Information

MSDS https://www.manchesterorganics.com/H14575
Availability details for
Quantity
  • Availability
  • Add to Cart