MAN326366875 Manchester Organics

4-Methyl-1,2-bis-(trifluoromethyl)benzene

  • MDL number MFCD14525508
  • Cas Number : 716-25-6
  • Molecular Weight : 228.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methyl-1,2-bis(trifluoromethyl)benzene
Inchi Code 1S/C9H6F6/c1-5-2-3-6(8(10,11)12)7(4-5)9(13,14)15/h2-4H,1H3
Inchi Key ZLZIWTFMKQRYNZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/U14514
Availability details for
Quantity
  • Availability
  • Add to Cart