MAN326366888 Manchester Organics

1-Methyl-4-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD14525512
  • Cas Number : 50562-01-1
  • Molecular Weight : 174.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-4-(2,2,2-trifluoroethyl)benzene
Inchi Code 1S/C9H9F3/c1-7-2-4-8(5-3-7)6-9(10,11)12/h2-5H,6H2,1H3
Inchi Key DSJJCEACYWFWAH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J14528
Availability details for
Quantity
  • Availability
  • Add to Cart