MAN326366892 Manchester Organics

1-Nitro-2-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD14525517
  • Cas Number : 145914-06-3
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/P14533
Availability details for
Quantity
  • Availability
  • Add to Cart