MAN326366895 Manchester Organics

1-Nitro-4-(2,2,2-trifluoro-1-methyl-ethyl)benzene

  • MDL number MFCD14525520
  • Cas Number : 1221272-94-1
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U14537
Availability details for
Quantity
  • Availability
  • Add to Cart