MAN326366923 Manchester Organics

2-Bromo-4-chlorotoluene

  • MDL number MFCD00060650
  • Cas Number : 27139-97-5
  • Molecular Weight : 205.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/U21827
Availability details for
Quantity
  • Availability
  • Add to Cart