MAN326366928 Manchester Organics

(1,1-Difluoropropyl)benzene

  • MDL number MFCD14525542
  • Cas Number : 74185-83-4
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R14559
Availability details for
Quantity
  • Availability
  • Add to Cart