MAN326366939 Manchester Organics

(2-Chloro-1,1-difluoroethyl)benzene

  • MDL number MFCD14525555
  • Cas Number : 55805-08-8
  • Molecular Weight : 176.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-1,1-difluoroethyl)benzene
Inchi Code 1S/C8H7ClF2/c9-6-8(10,11)7-4-2-1-3-5-7/h1-5H,6H2
Inchi Key RANLLZYBLOAACB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E14572
Availability details for
Quantity
  • Availability
  • Add to Cart