MAN326366944 Manchester Organics

1-(1,1-Difluoroethyl)-3-nitrobenzene

  • MDL number MFCD14525560
  • Cas Number : 40788-04-3
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1,1-difluoroethyl)-3-nitrobenzene
Inchi Code 1S/C8H7F2NO2/c1-8(9,10)6-3-2-4-7(5-6)11(12)13/h2-5H,1H3
Inchi Key QRIVJDGDHLGQAR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L14578
Availability details for
Quantity
  • Availability
  • Add to Cart