MAN326366958 Manchester Organics

1-Fluoro-4-(pentafluoroethyl)benzene

  • MDL number MFCD16140153
  • Molecular Weight : 214.108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X14588
Availability details for
Quantity
  • Availability
  • Add to Cart