MAN326366964 Manchester Organics

1-(Difluorophenylmethyl)-4-methylbenzene

  • MDL number MFCD16140157
  • Cas Number : 174075-29-7
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[difluoro(phenyl)methyl]-4-methylbenzene
Inchi Code 1S/C14H12F2/c1-11-7-9-13(10-8-11)14(15,16)12-5-3-2-4-6-12/h2-10H,1H3
Inchi Key XRPWOFFDNMCYGY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C14593
Availability details for
Quantity
  • Availability
  • Add to Cart