MAN326366991 Manchester Organics

6-Chloro-7-azaindole

  • MDL number MFCD06659666
  • Cas Number : 55052-27-2
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-3aH-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C7H5ClN2/c8-6-2-1-5-3-4-9-7(5)10-6/h1-4H,(H,9,10)
Inchi Key DSCOSIUAQUDGJM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T21849
Availability details for
Quantity
  • Availability
  • Add to Cart