MAN326366998 Manchester Organics

1-Chloro-2-(1,1-Difluoroethyl)benzene

  • MDL number MFCD14525553
  • Molecular Weight : 176.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C14618

Safety Information

MSDS https://www.manchesterorganics.com/C14618
Availability details for
Quantity
  • Availability
  • Add to Cart