MAN326367001 Manchester Organics

2-Fluoro-4-methoxyaniline

  • MDL number MFCD00047827
  • Cas Number : 458-52-6
  • Molecular Weight : 141.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/X21853
Availability details for
Quantity
  • Availability
  • Add to Cart