MAN326367028 Manchester Organics

2,6-Dichloro-3-nitrobenzotrifluoride

  • MDL number MFCD16140179
  • Molecular Weight : 259.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G14693

Safety Information

MSDS https://www.manchesterorganics.com/G14693
Availability details for
Quantity
  • Availability
  • Add to Cart