MAN326367035 Manchester Organics

3,5-Dichloro-4-(trifluoromethyl)pyridine

  • MDL number MFCD16140184
  • Cas Number : 1221272-92-9
  • Molecular Weight : 215.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T14632
Availability details for
Quantity
  • Availability
  • Add to Cart